6-Bromo-3-iodopyrazolo[1,5-a]pyrimidine 97%

Supplier: Ambeed
Warning

A284879-1G A284879-100MG A284879-250MG
103813-656EA 83 USD
103813-656 76837-320 76977-492 76977-494
6-Bromo-3-iodopyrazolo[1,5-a]pyrimidine 97%
6-Bromo-3-iodopyrazolo[1,5-a]pyrimidine

Precaución: For research use only

Formula: C₆H₃BrIN₃
MW: 323.9200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18073604
CAS Number: 1109284-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR