6-Bromo-3H-oxazolo[4,5-b]pyridin-2-one 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo[1,3]oxazolo[4,5-b]pyridin-2(3H)-one, 6-bromooxazolo[4,5-b]pyridin-2(3H)-one

A139879-25G A139879-1G A139879-250MG
103802-788EA 7 USD
103802-788 77299-098 77315-892 77685-196
6-Bromo-3H-oxazolo[4,5-b]pyridin-2-one 98%
6-Bromo-3H-oxazolo[4,5-b]pyridin-2-one

Precaución: for Research Use Only

Formula: C₆H₃BrN₂O₂
MW: 215.006 g/mol
Melting Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD08689590
CAS Number: 21594-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR