6-Bromo-3-fluoropicolinic acid 95%

Supplier: Ambeed
Warning

A592214-100MG A592214-250MG A592214-1G
103828-938EA 121 USD
103828-938 103840-084 76819-328 76819-330 76844-230
6-Bromo-3-fluoropicolinic acid 95%
6-Bromo-3-fluoropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185805
CAS Number: 1052714-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR