6-Bromo-3-chloropyrazin-2-amine 97%

Supplier: Ambeed
Warning

A358442-250MG A358442-100MG
103826-208EA 22 USD
103826-208 103837-214 77594-066 77672-802
6-Bromo-3-chloropyrazin-2-amine 97%
6-Bromo-3-chloropyrazin-2-amine

Precaución: Keep in a dark place, inert atmosphere, 2…8 °C

Formula: C₄H₃BrClN₃
MW: 208.4400 g/mol
Melting Pt: 147…151 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13193368
CAS Number: 1082843-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR