6-Bromo-1-tetralone 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Bromo-3,4-dihydro-1(2H)-naphthalenone, 6-Bromo-3,4-dihydronaphthalen-1(2H)-one
A254852-1G
A254852-5G
A254852-250MG
103812-172EA
25
USD
103812-172
103852-024
77091-476
77091-478
77166-060
6-Bromo-1-tetralone 97%
6-Bromo-1-tetralone
Precaución: for Research Use Only
Formula:
C₁₀H₉BrO MW: 225.08 g/mol Boiling Pt: 323.1 °C (760 mm Hg) Melting Pt: 40…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114378 CAS Number: 66361-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...