6-Bromo-3,4-dihydro-2H-1,4-benzoxazine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine

A199939-250MG A199939-1G A199939-25G A199939-5G
103817-150EA 24 USD
103817-150 103847-776 76814-234 77085-836 77085-838 77085-840
6-Bromo-3,4-dihydro-2H-1,4-benzoxazine 98%
6-Bromo-3,4-dihydro-2H-1,4-benzoxazine

Precaución: for Research Use Only

Formula: C₈H₈BrNO
MW: 214.062 g/mol
Boiling Pt: 296.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08544341
CAS Number: 105655-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR