6-Bromo-3,4-dihydro-1H-quinolin-2-one 98%

Supplier: Ambeed
Warning

A240802-100G A240802-10G A240802-25G A240802-5G A240802-1G
103811-364EA 11 USD
103811-364 103851-170 77000-422 77000-424 77090-324 77090-326 77304-202
6-Bromo-3,4-dihydro-1H-quinolin-2-one 98%
6-Bromo-3,4-dihydro-1H-quinolin-2-one

Precaución: for Research Use Only

Formula: C₉H₈BrNO
Boiling Pt: 376 °C (760 mm Hg)
Flash Pt: 181.4 °C
Storage Temperature: Ambient
MDL Number: MFCD03839832
CAS Number: 3279-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR