6-Bromo-3,4-dihydro-1H-[1,8]naphthyridin-2-one 95%

Supplier: Ambeed
Warning

A177642-10G A177642-250MG A177642-1G A177642-25G A177642-5G
103807-256EA 23 USD
103807-256 103845-588 77011-830 77011-832 77082-974 77082-976 77082-978
6-Bromo-3,4-dihydro-1H-[1,8]naphthyridin-2-one 95%
6-Bromo-3,4-dihydro-1H-[1,8]naphthyridin-2-one

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O
MW: 227.0600 g/mol
Boiling Pt: 398.2 °C (760 mm Hg)
Melting Pt: 265…267 °C
Storage Temperature: Ambient
MDL Number: MFCD05664027
CAS Number: 129686-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR