6-Bromo-3-(trifluoromethyl)-1H-indazole 95%

Supplier: Ambeed
Warning

A171084-100MG A171084-250MG A171084-1G A171084-5G
103806-586EA 142 USD
103806-586 103844-860 76985-418 76985-420 77081-982 77081-984
6-Bromo-3-(trifluoromethyl)-1H-indazole 95%
6-Bromo-3-(trifluoromethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₄BrF₃N₂
MW: 265.03 g/mol
Boiling Pt: 328.2±37.0 °C (760 mm Hg)
Flash Pt: 152.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD26685102
CAS Number: 1374258-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR