6-Bromo-2-phenylimidazo[1,2-a]pyridine 97%

Supplier: Ambeed
Warning

A140064-10G A140064-250MG A140064-1G
103802-826EA 12 USD
103802-826 77027-462 77027-464 77315-932
6-Bromo-2-phenylimidazo[1,2-a]pyridine 97%
6-Bromo-2-phenylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₁₃H₉BrN₂
MW: 273.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00219253
CAS Number: 4044-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR