6-Bromo-2-naphthylmethanol 97%

Supplier: Ambeed
Warning

A226129-100MG A226129-250MG A226129-1G A226129-5G
103819-226EA 43 USD
103819-226 103850-126 77030-942 77030-944 77088-854 77088-856
6-Bromo-2-naphthylmethanol 97%
6-Bromo-2-naphthylmethanol

Precaución: for Research Use Only

Formula: C₁₁H₉BrO
MW: 237.09 g/mol
Boiling Pt: 374.611 °C (760 mm Hg)
Melting Pt: 152…153 °C
Flash Pt: 180.359 °C
Storage Temperature: Ambient
MDL Number: MFCD09842470
CAS Number: 100751-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR