6-Bromo-2-naphthol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Bromo-2-hydroxynaphthalene, 2-Bromo-6-hydroxynaphthalene
A190680-1000G
A190680-100G
A190680-10G
A190680-500G
A190680-25G
A190680-5G
103846-920EA
5
USD
103846-920
76998-962
76998-964
76998-966
76998-968
77084-730
77084-732
6-Bromo-2-naphthol 98%
6-Bromo-2-naphthol
Precaución: for Research Use Only
Formula:
C₁₀H₇BrO Melting Pt: 122…124 °C Flash Pt: 167.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004081 CAS Number: 15231-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...