6-Bromo-2-naphthoic acid 98%
Supplier: Ambeed
|
Warning
|
A238794-100G
A238794-10G
A238794-5G
A238794-25G
103851-040EA
13
USD
103851-040
76801-294
76801-296
77090-174
77304-120
6-Bromo-2-naphthoic acid 98%
6-Bromo-2-naphthoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₇BrO₂ MW: 251.08 g/mol Boiling Pt: 387.3 °C (760 mm Hg) Melting Pt: 294…295 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075720 CAS Number: 5773-80-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...