6-Bromo-2-methylquinazolin-4(3H)-one 98%

Supplier: Ambeed
Warning

A151777-250MG A151777-1G A151777-5G A151777-10G
103804-300EA 30 USD
103804-300 103842-450 76768-166 77318-034 77318-036 77624-874
6-Bromo-2-methylquinazolin-4(3H)-one 98%
6-Bromo-2-methylquinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O
Boiling Pt: 372.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00548197
CAS Number: 5426-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR