2-Bromo-5-cyano-6-picoline 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2-methylnicotinonitrile

A162228-100MG A162228-250MG A162228-5G
103805-592EA 99 USD
103805-592 103843-842 76872-070 76872-072 77300-580
2-Bromo-5-cyano-6-picoline 97%
2-Bromo-5-cyano-6-picoline

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.032 g/mol
Boiling Pt: 267.5±35.0 °C (760 mm Hg)
Flash Pt: 115.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09839266
CAS Number: 1003711-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR