6-Bromo-2-methylisoquinolin-1(2H)-one 95%

Supplier: Ambeed
Warning

A184392-100MG A184392-250MG A184392-1G
103815-752EA 95 USD
103815-752 103846-314 76784-694 76784-696 77083-926
6-Bromo-2-methylisoquinolin-1(2H)-one 95%
6-Bromo-2-methylisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
Flash Pt: 179.4 °C
Storage Temperature: Ambient
MDL Number: MFCD11858430
CAS Number: 864866-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR