6-Bromo-2-methylisoindolin-1-one 97%

Supplier: Ambeed
Warning

A153276-100MG A153276-250MG A153276-5G
103804-488EA 61 USD
103804-488 103842-660 77010-906 77010-908 77318-310
6-Bromo-2-methylisoindolin-1-one 97%
6-Bromo-2-methylisoindolin-1-one

Precaución: for Research Use Only

Formula: C₉H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD17676936
CAS Number: 1254319-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR