6-Bromo-2-methyl-3-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A407811-1G A407811-100MG A407811-250MG
103827-162EA 235 USD
103827-162 76841-696 77240-708 77240-710
6-Bromo-2-methyl-3-nitrobenzoic acid 97%
6-Bromo-2-methyl-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrNO₄
Storage Temperature: Ambient
CAS Number: 1207341-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR