6-Bromo-2-methyl-2H-indazole 97%
Supplier: Ambeed
|
Warning
|
A156535-10G
A156535-250MG
A156535-5G
A156535-1G
A156535-25G
103804-902EA
16
USD
103804-902
103843-100
76783-614
76783-616
77300-194
77318-890
77318-892
6-Bromo-2-methyl-2H-indazole 97%
6-Bromo-2-methyl-2H-indazole
Precaución: for Research Use Only
Formula:
C₈H₇BrN₂ MW: 211.0600 g/mol Boiling Pt: 322.7 °C (760 mm Hg) Melting Pt: 82…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD09870048 CAS Number: 590417-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...