6-Bromo-2-methyl-[1,2,4]triazolo[1,5-a]pyridine 98%

Supplier: Ambeed
Warning

A156832-100MG A156832-250MG A156832-1G A156832-5G
103804-926EA 77 USD
103804-926 103843-130 76768-388 76768-390 77318-932 77318-934
6-Bromo-2-methyl-[1,2,4]triazolo[1,5-a]pyridine 98%
6-Bromo-2-methyl-[1,2,4]triazolo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
Melting Pt: 153…155 °C
Storage Temperature: Ambient
MDL Number: MFCD09038167
CAS Number: 7169-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR