3-Amino-6-bromo-2-methoxypyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Bromo-2-methoxypyridin-3-amine
A101935-10G
A101935-250MG
A101935-25G
A101935-1G
A101935-5G
103796-206EA
9
USD
103796-206
103821-904
77148-244
77148-246
77148-248
77308-794
77308-796
3-Amino-6-bromo-2-methoxypyridine 97%
3-Amino-6-bromo-2-methoxypyridine
Precaución: for Research Use Only
Formula:
C₆H₇BrN₂O Boiling Pt: 277.6 °C (760 mm Hg) Melting Pt: 76…80 °C Flash Pt: 121.683 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06738312 CAS Number: 89466-18-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...