6-Bromo-2-methoxy-3-(4-methyl-1H-imidazol-1-yl)pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 1-(6-Bromo-2-methoxypyridin-3-yl)-4-methyl-1H-imidazole

A204485-100MG A204485-250MG A204485-25G A204485-1G A204485-5G
103817-556EA 72 USD
103817-556 103848-220 77152-272 77152-274 77152-276 77302-912 77302-914
6-Bromo-2-methoxy-3-(4-methyl-1H-imidazol-1-yl)pyridine 95%
6-Bromo-2-methoxy-3-(4-methyl-1H-imidazol-1-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrN₃O
MW: 268.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19690830
CAS Number: 1123194-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR