6-Bromo-2H-1,4-benzoxazin-3(4H)-one 97%

Supplier: Ambeed
Warning

A144804-100G A144804-10G A144804-250MG A144804-25G A144804-5G A144804-1G
103803-450EA 10 USD
103803-450 103841-528 76767-960 76767-962 76767-964 77299-410 77299-412 77316-780
6-Bromo-2H-1,4-benzoxazin-3(4H)-one 97%
6-Bromo-2H-1,4-benzoxazin-3(4H)-one

Precaución: for Research Use Only

Formula: C₈H₆BrNO₂
Boiling Pt: 376.8 °C (760 mm Hg)
Melting Pt: 220…225 °C
Flash Pt: 181.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00461173
CAS Number: 24036-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR