6-Bromo-2-chloroquinoxaline 98%

Supplier: Ambeed
Warning

A129412-100MG A129412-250MG A129412-1G
103801-280EA 45 USD
103801-280 76782-240 76782-242 77313-664
6-Bromo-2-chloroquinoxaline 98%
6-Bromo-2-chloroquinoxaline

Precaución: for Research Use Only

Formula: C₈H₄BrClN₂
Boiling Pt: 312.5 °C (760 mm Hg)
Melting Pt: 150…152.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09260998
CAS Number: 55687-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR