6-Bromo-2-chloroquinoline 98%

Supplier: Ambeed
Warning

A286556-1G A286556-25G A286556-5G A286556-100G A286556-10G A286556-250MG
103813-722EA 9 USD
103813-722 103834-858 76837-396 76837-398 76837-400 77262-726 77262-728 77262-730
6-Bromo-2-chloroquinoline 98%
6-Bromo-2-chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
MW: 242.50 g/mol
Boiling Pt: 325.7 °C (760 mm Hg)
Melting Pt: 148…150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115272
CAS Number: 1810-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR