6-Bromo-2-chloropyridin-3-amine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Amino-6-bromo-2-chloropyridine
A200503-100G
A200503-10G
A200503-250MG
A200503-1G
A200503-25G
A200503-5G
103817-212EA
6
USD
103817-212
103847-840
77030-112
77030-114
77030-116
77085-904
77085-906
77085-908
6-Bromo-2-chloropyridin-3-amine 97%
6-Bromo-2-chloropyridin-3-amine
Precaución: for Research Use Only
Formula:
C₅H₄BrClN₂ MW: 207.4600 g/mol Boiling Pt: 302.4 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD04971930 CAS Number: 169833-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...