6-Bromo-2,4-dichloroquinazoline 97%

Supplier: Ambeed
Warning

A129792-250MG A129792-1G A129792-10G
103801-326EA 14 USD
103801-326 77026-998 77313-714 77566-520
6-Bromo-2,4-dichloroquinazoline 97%
6-Bromo-2,4-dichloroquinazoline

Precaución: For research use only.

Formula: C₈H₃BrCl₂N₂
MW: 277.93 g/mol
Boiling Pt: 330.1 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD09744007
CAS Number: 102393-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR