6-Bromo-2,3-dihydroquinolin-4(1H)-one 95%

Supplier: Ambeed
Warning

A349800-100MG A349800-250MG A349800-1G A349800-5G
103826-026EA 77 USD
103826-026 103837-020 76788-370 76788-372 77306-604 77306-606
6-Bromo-2,3-dihydroquinolin-4(1H)-one 95%
6-Bromo-2,3-dihydroquinolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₉H₈BrNO
MW: 226.0700 g/mol
Boiling Pt: 359.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00600620
CAS Number: 76228-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR