6-Bromo-2,3-dihydro-1H-indene-1-carbonitrile 97%

Supplier: Ambeed
Warning

A126716-100MG A126716-250MG A126716-5G
103800-854EA 59 USD
103800-854 77149-448 77149-450 77298-144
6-Bromo-2,3-dihydro-1H-indene-1-carbonitrile 97%
6-Bromo-2,3-dihydro-1H-indene-1-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MW: 222.08 g/mol
Melting Pt: 65…66 °C
Storage Temperature: Ambient
MDL Number: MFCD11219697
CAS Number: 783335-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR