6-Bromo-2',3',5',6'-tetrahydrospiro[indoline-3,4'-pyran]-2-one 97%

Supplier: Ambeed
Warning

A100617-100MG A100617-250MG A100617-1G A100617-5G
103795-540EA 175 USD
103795-540 103821-230 76982-138 76982-140 77282-524 77282-526
6-Bromo-2',3',5',6'-tetrahydrospiro[indoline-3,4'-pyran]-2-one 97%
6-Bromo-2',3',5',6'-tetrahydrospiro[indoline-3,4'-pyran]-2-one

Precaución: for Research Use Only

Formula: C₁₂H₁₂BrNO₂
Boiling Pt: 437.1±45.0 °C (760 mm Hg)
Flash Pt: 218.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19703313
CAS Number: 1190861-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR