6-Bromo-1-methylpyridin-2(1H)-one 97%

Supplier: Ambeed
Warning

A112089-100MG A112089-10G A112089-250MG A112089-1G
103797-952EA 65 USD
103797-952 103823-786 76809-976 76809-978 76809-980 77310-596
6-Bromo-1-methylpyridin-2(1H)-one 97%
6-Bromo-1-methylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₆BrNO
Boiling Pt: 287.1±33.0 °C (760 mm Hg)
Flash Pt: 127.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00234174
CAS Number: 873383-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR