6-Bromo-1-methyl-1H-pyrazolo[4,3-b]pyridine 95%

Supplier: Ambeed
Warning

A213097-1G A213097-100MG
103818-260EA 127 USD
103818-260 77303-270 77671-900
6-Bromo-1-methyl-1H-pyrazolo[4,3-b]pyridine 95%
6-Bromo-1-methyl-1H-pyrazolo[4,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
Boiling Pt: 287.0±20.0 °C (760 mm Hg)
Flash Pt: 127.4 °C
Storage Temperature: Ambient
MDL Number: MFCD12024535
CAS Number: 1150617-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR