3-Amino-6-bromo-1-methyl-1H-indazole 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-1-methyl-1H-indazol-3-amine

A159213-100MG A159213-250MG A159213-1G A159213-25G
103805-232EA 17 USD
103805-232 103843-448 76783-768 76783-770 77300-390 77550-000
3-Amino-6-bromo-1-methyl-1H-indazole 97%
3-Amino-6-bromo-1-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₈BrN₃
MW: 226.0700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847108
CAS Number: 1214899-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR