6-Bromo-1-methyl-1H-benzo[d][1,2,3]triazole 97%

Supplier: Ambeed
Warning

A121654-100MG A121654-250MG A121654-1G A121654-5G
103800-104EA 31 USD
103800-104 76971-548 76971-550 77297-842 77312-230
6-Bromo-1-methyl-1H-benzo[d][1,2,3]triazole 97%
6-Bromo-1-methyl-1H-benzo[d][1,2,3]triazole

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.047 g/mol
Boiling Pt: 268 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11559050
CAS Number: 1083181-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR