6-Bromo-1-indanone 97% (by HPLC)

Supplier: Ambeed
Warning

A165367-10G A165367-1G A165367-5G
103805-960EA 5 USD
103805-960 103844-216 77151-050 77300-778 77320-418
6-Bromo-1-indanone 97% (by HPLC)
6-Bromo-1-indanone

Precaución: for Research Use Only

Formula: C₉H₇BrO
MW: 211.06 g/mol
Melting Pt: 111…115 °C
Flash Pt: 111.6 °C
Storage Temperature: Ambient
MDL Number: MFCD02179286
CAS Number: 14548-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR