6-Bromo-1H-pyrrolo[3,2-b]pyridine-3-carbaldehyde 98%

Supplier: Ambeed
Warning

A116574-250MG A116574-1G
103798-756EA 61 USD
103798-756 103824-620 76810-216 77297-474
6-Bromo-1H-pyrrolo[3,2-b]pyridine-3-carbaldehyde 98%
6-Bromo-1H-pyrrolo[3,2-b]pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O
MW: 225.042 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12963037
CAS Number: 1190312-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR