6-Bromo-1H-pyrrolo[3,2-b]pyridine 97%

Supplier: Ambeed
Warning

A181820-100G A181820-10G A181820-250MG A181820-1G A181820-25G A181820-5G
103807-656EA 20 USD
103807-656 103846-028 76769-576 76769-578 76769-580 77083-552 77083-554 77083-556
6-Bromo-1H-pyrrolo[3,2-b]pyridine 97%
6-Bromo-1H-pyrrolo[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.0300 g/mol
Boiling Pt: 311 °C (760 mm Hg)
Flash Pt: 142 °C
Storage Temperature: Ambient
MDL Number: MFCD09266227
CAS Number: 944937-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR