6-Bromo-1H-pyrazolo[4,3-c]pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-5-azaindazole

A250731-10G A250731-250MG A250731-1G A250731-25G A250731-5G
103811-948EA 46 USD
103811-948 103851-786 76815-644 76815-646 77091-148 77091-150 77091-152
6-Bromo-1H-pyrazolo[4,3-c]pyridine 97%
6-Bromo-1H-pyrazolo[4,3-c]pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MW: 198.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17010106
CAS Number: 1206973-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR