6-Bromo-1H-indole-2-carboxylic acid 96%

Supplier: Ambeed
Warning

A122895-10G A122895-250MG A122895-1G A122895-5G A122895-25G
103800-302EA 19 USD
103800-302 77271-612 77271-614 77297-898 77312-424 77571-398
6-Bromo-1H-indole-2-carboxylic acid 96%
6-Bromo-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
Melting Pt: 221…223 °C
Storage Temperature: Ambient
MDL Number: MFCD02664469
CAS Number: 16732-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR