6-Bromo-1H-indole-2-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-1H-indole-2-carboxaldehyde

A633939-100MG A633939-250MG A633939-5G
103840-298EA 401 USD
103840-298 77018-238 77018-240 77308-018
6-Bromo-1H-indole-2-carbaldehyde 98%
6-Bromo-1H-indole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MW: 224.06 g/mol
Boiling Pt: 395.6 °C (760 mm Hg)
Melting Pt: 181…183 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06738308
CAS Number: 105191-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR