6-Bromo-1H-indazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A131993-250MG A131993-1G A131993-10G
103801-672EA 15 USD
103801-672 76782-398 77298-530 77871-615
6-Bromo-1H-indazole-4-carboxylic acid 97%
6-Bromo-1H-indazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O₂
Melting Pt: 293…298 °C
Storage Temperature: Ambient
MDL Number: MFCD07368215
CAS Number: 885523-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR