6-Bromo-1H-indazole-3-carbaldehyde 98%

Supplier: Ambeed
Warning

A137964-10G A137964-250MG A137964-1G A137964-5G
103802-534EA 66 USD
103802-534 77149-936 77149-938 77315-532 77315-534
6-Bromo-1H-indazole-3-carbaldehyde 98%
6-Bromo-1H-indazole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O
MW: 225.042 g/mol
Boiling Pt: 414.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06738286
CAS Number: 885271-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR