6-Bromo-1H-indazole 98%
Supplier: Ambeed
|
Warning
|
A109835-100G
A109835-10G
A109835-25G
A109835-500G
A109835-5G
A109835-1G
103797-574EA
9
USD
103797-574
103823-378
77160-662
77160-664
77160-666
77160-668
77283-892
77310-184
6-Bromo-1H-indazole 98%
6-Bromo-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₅BrN₂ MW: 197.0300 g/mol Melting Pt: 180…185 °C Flash Pt: 238.611 °C Storage Temperature: Ambient |
MDL Number:
MFCD03265457 CAS Number: 79762-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...