6-Bromo-1H-indazol-3-ol 98%

Supplier: Ambeed
Warning

A347526-1G A347526-5G A347526-250MG
103825-970EA 20 USD
103825-970 103836-956 76840-036 77306-584 77878-056
6-Bromo-1H-indazol-3-ol 98%
6-Bromo-1H-indazol-3-ol

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD07781600
CAS Number: 885521-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR