6-Bromo-1-hexene 98%

Supplier: Ambeed
Danger

A251046-25G A251046-1G A251046-10G
103851-800EA 6 USD
103851-800 77091-168 77098-686 77568-434
6-Bromo-1-hexene 98%
6-Bromo-1-hexene

Precaución: For research use only.

Formula: C₆H₁₁Br
MW: 163.06 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00000269
CAS Number: 2695-47-8
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR