6-Bromo-1H-benzo[d]imidazole-4-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A233253-250MG
A233253-100MG
103810-876EA
163
USD
103810-876
103850-636
77600-872
77626-234
6-Bromo-1H-benzo[d]imidazole-4-carboxylic acid 98%
6-Bromo-1H-benzo[d]imidazole-4-carboxylic acid
Precaución: Sealed in dry, at room temperature.
Formula:
C₈H₅BrN₂O₂ MW: 241.04 g/mol Boiling Pt: 576.2 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD06411035 CAS Number: 255064-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...