6-Bromo-1-chloroisoquinoline 97%

Supplier: Ambeed
Warning

A112852-100G A112852-10G A112852-250MG A112852-25G A112852-1G A112852-5G
103798-120EA 14 USD
103798-120 103823-956 77009-002 77009-004 77009-006 77009-008 77310-784 77310-786
6-Bromo-1-chloroisoquinoline 97%
6-Bromo-1-chloroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
Boiling Pt: 349.5 °C (760 mm Hg)
Flash Pt: 165.17 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06738661
CAS Number: 205055-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR