6-Bromo-1,3-dimethyl-1H-indazole 97%

Supplier: Ambeed
Warning

A221074-1G A221074-100MG A221074-250MG A221074-5G
103818-888EA 63 USD
103818-888 103849-768 77088-362 77165-214 77165-216 77303-576
6-Bromo-1,3-dimethyl-1H-indazole 97%
6-Bromo-1,3-dimethyl-1H-indazole

Precaución: for Research Use Only

Formula: C₉H₉BrN₂
MW: 225.085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12028626
CAS Number: 1095539-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR