6-Bromo-1,2,3,4-tetrahydroisoquinoline 95%

Supplier: Ambeed
Warning

A306786-1G A306786-5G A306786-250MG A306786-10G
103814-482EA 19 USD
103814-482 103835-654 76838-382 76838-384 77015-076 77568-616
6-Bromo-1,2,3,4-tetrahydroisoquinoline 95%
6-Bromo-1,2,3,4-tetrahydroisoquinoline

Precaución: For research use only.

Formula: C₉H₁₀BrN
Boiling Pt: 283 °C (760 mm Hg)
Flash Pt: 125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07374370
CAS Number: 226942-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR