6-Bromo-1-(tetrahydro-2H-pyran-4-yl)-1H-benzo[d]imidazole 98%

Supplier: Ambeed
Warning

A116653-100MG A116653-250MG A116653-25G A116653-1G A116653-5G
103798-780EA 89 USD
103798-780 103824-644 77026-156 77026-158 77026-160 77297-486 77297-488
6-Bromo-1-(tetrahydro-2H-pyran-4-yl)-1H-benzo[d]imidazole 98%
6-Bromo-1-(tetrahydro-2H-pyran-4-yl)-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrN₂O
MW: 281.14 g/mol
Boiling Pt: 425.3±55.0 °C (760 mm Hg)
Flash Pt: 211 °C
Storage Temperature: Ambient
MDL Number: MFCD18072651
CAS Number: 1245649-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR